Learn More
Fisherbrand™ Adapters for Fisherbrand™ accuSpin™ 400 6-Place Fixed-Angle Rotor
Brand:  Fisherbrand™
Code : 51
Additional Details : Weight : 0.07920kg
Product Code. 10658612
  • £77.10 / Each
In Stock
to see stock.
Description
Description
For use with fixed-angle rotor 13-100-559.
Adapters are sold individually. Order the correct quantity to outfit rotor.
Specifications
Specifications
Conical
400 6-place Fixed angle rotor
15 mL