missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Adapters For 100mL Rectangular Bucket
For use with Eppendorf Tubes
Brand: Eppendorf™ 5804751000
131.00 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Adapters for rectangular buckets
- For use with Rotor A-4-44
- Available in a variety of sizes for a wide range of applications
Specifications
Adapter | |
100mL Rectangular Bucket | |
48 |
1.5 to 2 mL | |
11 mm | |
2 Pack |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction