missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Adapters For 400mL Bucket

Use these adaptors with 400mL buckets for Eppendorf™ rotors.

£156.00 - £192.77

Specifications

Type Adapter
No. per Pack 2 Pack
View More Specs

Products 4
Product Code Brand Capacity Tube Type Price Quantity & Availability  
Product Code Brand Capacity Tube Type Price Quantity & Availability  
11850252
View accessory products View Documents
Eppendorf™
5810728002
400 mL Flat
£156.00
Pack of 2
Please to purchase this item. Need a web account? Register with us today!
11578683
View Documents
Eppendorf™
5810721008
7 to 17 mL Flat
£182.73
Pack of 2
Please to purchase this item. Need a web account? Register with us today!
11588623
View Documents Promotion Details
Eppendorf™
5810720001
2.6 to 5 mL Flat
£192.77
Pack of 2
Please to purchase this item. Need a web account? Register with us today!
10263592
View Documents Promotion Details
Eppendorf™
5810723000
50 mL Conical
£192.77
Pack of 2
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Two adapters per pack
  • Compatible with conical or flat tubes and buckets for Eppendorf rotors
Specifications

Specifications

Adapter
2 Pack
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Eppendorf™ Adapters For 400mL Bucket

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.