Learn More
Thermo Scientific™ Adapters for Thermo Scientific™ MicroClick 30 x 2 Rotor and MicroClick 24 x 2 Rotor
PCR Tube and Microtube Adapters
Brand: Thermo Scientific™ 75005753
221.80 GBP valid until 2024-11-30
Use promo code "21681" to get your promotional price.
This item is not returnable.
View return policy
Description
Adapters for Thermo Scientific MicroClick 30 x 2 Rotor and MicroClick 24 x 2 Rotor
Specifications
Rotor Adapter | |
0.5 mL | |
0.5 mL |
Microtube | |
MicroClick 30 x 2 and MicroClick 24 x 2 rotor |