missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva ADCF-MAb Media
Increase process yields for the manufacture of monoclonal antibodies (MAbs) with HyClone™ ADCF-MAb Media, a protein-free media containing no animal-derived components.
£31.55 - £445.00
Specifications
Certifications/Compliance | cGMP |
---|---|
Content And Storage | 2° to 8°C, away from light |
Description | with L-Glutamine |
Form | Liquid |
Product Type | ADCF-mAb Liquid Medium |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
10788134
|
Cytiva
SH30349.01 |
with L-Glutamine | 500 mL bottle |
£31.55
500mL |
Estimated Shipment: 06-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10094723
|
Cytiva
SH30349.LS |
with L-Glutamine | Six 1000 mL bottles |
£445.00
Pack of 6 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10397042
|
Cytiva
SH30349.02 |
with L-Glutamine | 1000 mL bottle |
£68.05
1000mL |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- ADCF
- Metabolically designed for high cell yield and monoclonal antibody production
- Allows for direct or sequential adaptation
- Component traceability
- Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
- Contains L-glutamine and Poloxamer 188
- Formulations without L-glutamine support the GS gene expression system
Specifications
cGMP | |
with L-Glutamine | |
ADCF-mAb Liquid Medium | |
Glutamine, Poloxamer 188, HEPES, Sodium Bicarbonate |
2° to 8°C, away from light | |
Liquid | |
0.2 μm Sterile-filtered | |
No Protein, No Phenol Red |