missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Additional Humidity Tray

For Use With: Excella ECO-170 and Innova CO-170 Incubators

Brand:  Eppendorf™ P0628-6260

Product Code. 11319954

  • £873.48 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • For 170 L incubators
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2.5 L
Accessory, Tray
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers