Learn More
Techne™ Additional Loading Dock
Techne™ Additional Loading Dock for use with the Prime Pro 48 Real Time PCR System
Brand: Techne™ 99966-70
Code : NEW
Additional Details : Weight : 0.50000kg
Description
- This ergonomic plate loading dock allows for easy loading of a plate
- LED Backlight allows the user to easily visualise sample loading
- Plate adapter fits standard centrifuge rotors
Specifications
2 Centrifuge Adapters | |
Prime Pro 48 Real-Time PCR System | |
Additional Loading Dock |