Learn More
BINDER™ Additional Universal Ventilation
Additional universal connection for ventilation with ambient air or inert gas. Flow limiter thread (RP in.), and connection with 3/8 in. male thread.
Brand: BINDER™ 8612-0001
Additional Details : Weight : 0.10000kg
Description
- 3/8 in. male thread connection or hose with 8 mm internal diameter
- The additional connection is made of stainless steel
Specifications
Additional universal ventilation - Additional universal connection for ventilation with ambient air or inert gas. Flow limiter thread (RP in.), and connection with 3/8 in. male thread. | |
Access Port | |
Universal |
BINDER Chambers Series VD, VDL | |
Stainless Steel | |
120/230 V |