missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
CAS | 73-24-5 |
---|---|
Molecular Formula | C5H5N5 |
Molecular Weight (g/mol) | 135.13 |
MDL Number | MFCD00041790 |
InChI Key | GFFGJBXGBJISGV-UHFFFAOYSA-N |
Synonym | adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine |
PubChem CID | 190 |
ChEBI | CHEBI:16708 |
IUPAC Name | 7H-purin-6-amine |
SMILES | NC1=C2NC=NC2=NC=N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10716181
|
Thermo Scientific Chemicals
147440250 |
25 g | Glass bottle |
£86.13
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10746341
|
Thermo Scientific Chemicals
147441000 |
100 g | Glass bottle |
£235.17
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10444595
|
Thermo Scientific Chemicals
147445000 |
500 g | Glass bottle |
£794.96
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
73-24-5 | |
135.13 | |
GFFGJBXGBJISGV-UHFFFAOYSA-N | |
190 | |
7H-purin-6-amine |
C5H5N5 | |
MFCD00041790 | |
adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine | |
CHEBI:16708 | |
NC1=C2NC=NC2=NC=N1 |
Specifications
73-24-5 | |
99.5 | |
360°C to 365°C | |
0.5% max. (105°C, 5 hrs) | |
98.5% min. (HPLC) | |
MFCD00041790 | |
15, 140 | |
Solubility in water: 0.5g/L (20°C). Other solubilities: soluble in acids and alkalies,slightly soluble in ethanol,insoluble in chloroform and ether,25g/l in boiling water | |
98.0 to 102.0% (lambda max. 262nm, pH 1), molar absorptivity 13100 +/- 2%, on dry basis | |
NC1=C2NC=NC2=NC=N1 | |
135.13 | |
CHEBI:16708 | |
99% | |
Adenine |
0.74 to 0.78 (A250/A260), 0.36 to 0.4 (A280/A260) | |
100.0 | |
Colorless to Yellow | |
Authentic | |
C5H5N5 | |
26, 420 | |
adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine | |
220°C | |
GFFGJBXGBJISGV-UHFFFAOYSA-N | |
7H-purin-6-amine | |
190 | |
135.13 | |
Liquid or Solid |
Safety and Handling
GHS H Statement
Toxic if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-796-1
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Adenine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only