missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenine, 99%

Product Code. 11304619
Click to view available options
Quantity:
25 g
50 g
250 g
500 g
Unit Size:
250g
25g
500g
50g
This item is not returnable. View return policy

Product Code. 11304619

Brand: Thermo Scientific Chemicals A14906.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 73-24-5 | C5H5N5 | 135.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Adenine is used as an active component of boron-deficient media to grow yeast in order to assess whether yeast growth is stimulated by boron. It is useful as a local antiseptic and vitamin B4. Further, it is used in the microbial determination of niacin. It is also employed as a food supplement for adult rats to investigate the effects of dietary adenine overload. In addition to this, it is used in the production of nucleotides of the nucleic acids.

Solubility
Soluble in hot water, methanol and ethanol. Slightly soluble in cold water. Insoluble in diethyl ether.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 73-24-5
Molecular Formula C5H5N5
Molecular Weight (g/mol) 135.13
MDL Number MFCD00041790
InChI Key GFFGJBXGBJISGV-UHFFFAOYSA-N
Synonym adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine
PubChem CID 190
ChEBI CHEBI:16708
IUPAC Name 7H-purin-6-amine
SMILES NC1=C2NC=NC2=NC=N1

Specifica

Melting Point >360°C (decomposition)
Assay Percent Range 99%
Odor Odorless
Quantity 50 g
UN Number UN2811
Beilstein 5777
Merck Index 14,152
Solubility Information Soluble in hot water,methanol and ethanol. Slightly soluble in cold water. Insoluble in diethyl ether.
Formula Weight 135.13
Percent Purity 99%
Chemical Name or Material Adenine
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • Adenine
Avvertenza
  • Danger
Categoria di pericolo
  • Acute toxicity Category 3
Indicazioni di pericolo
  • H301-Toxic if swallowed.
Consigli di prudenza
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Informazioni supplementari
  • MIXTURE LIST-Contains : Adenine

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato