Learn More
Tocris Bioscience™ Adenosine
Endogenous adenosine receptor agonist
Brand:  Tocris Bioscience™ 3624/50
Product Code. 18004642
  • £90.00 / 50mg
Explore more special offers
Description
Description
Adenosine is a neurotransmitter that acts as the preferred endogenous agonist at all adenosine receptor subtypes.
Specifications
Specifications
>99%
Adenosine
Adenosine
50 mg
267.24