missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine 5'-monophosphate sodium salt hydrate, 99%, Thermo Scientific Chemicals
£20.50 - £167.00
Chemical Identifiers
CAS | 149022-20-8 |
---|---|
Molecular Formula | C10H13N5NaO7P |
Molecular Weight (g/mol) | 369.21 |
MDL Number | MFCD18785650 |
InChI Key | FYWLYWMEGHCZAX-GWKNMROSNA-M |
Synonym | sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate x:1:x, adenosine 5-monophosphoric acid disodium salt, adenosine 5'-monophosphate sodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate, pubchem14181, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate 2:1:x, 5'-adenylic acid,sodium salt, hydrate 9ci, disodium hydrate adenosine-5-monophosphate 2- |
PubChem CID | 23674992 |
IUPAC Name | sodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate |
SMILES | [Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11986851
|
Thermo Scientific Acros
440710010 |
1 g | Glass bottle |
£20.50
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11996851
|
Thermo Scientific Acros
440710050 |
5 g | Glass bottle |
£47.13
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11906861
|
Thermo Scientific Acros
440710250 |
25 g | Glass bottle |
£167.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
149022-20-8 | |
369.21 | |
FYWLYWMEGHCZAX-GWKNMROSNA-M | |
23674992 | |
[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1 |
C10H13N5NaO7P | |
MFCD18785650 | |
sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate x:1:x, adenosine 5-monophosphoric acid disodium salt, adenosine 5'-monophosphate sodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate, pubchem14181, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate 2:1:x, 5'-adenylic acid,sodium salt, hydrate 9ci, disodium hydrate adenosine-5-monophosphate 2- | |
sodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate |
Specifications
149022-20-8 | |
98.5 | |
4.50 to 6.50 | |
98.5% min. (HPLC) | |
MFCD18785650 | |
FYWLYWMEGHCZAX-GWKNMROSNA-M | |
sodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate | |
23674992 | |
99% | |
Adenosine 5′-monophosphate sodium salt hydrate |
White | |
100.0 | |
Powder | |
C10H13N5NaO7P | |
sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate x:1:x, adenosine 5-monophosphoric acid disodium salt, adenosine 5'-monophosphate sodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate, pubchem14181, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate 2:1:x, 5'-adenylic acid,sodium salt, hydrate 9ci, disodium hydrate adenosine-5-monophosphate 2- | |
[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1 | |
369.21 | |
369.21 | |
Authentic |
RUO – Research Use Only