missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Adenosine-5'-triphosphate disodium salt hydrate, 98% Product Code.: 15450177

Thermo Scientific Chemicals Adenosine-5'-triphosphate disodium salt hydrate, 98%

Código de producto. 15450177
5 g
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
10 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones

Product Code. 15450177

Brand: Thermo Scientific Chemicals J61125.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Adenosine-5'-triphosphate disodium salt hydrate, is used as a P2 purinoceptor agonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenosine-5′-triphosphate disodium salt hydrate, is used as a P2 purinoceptor agonist.

Solubility
Soluble in water (110 mg/ml at 25°C), ethanol (<1mg/mL at 25°C), DMSO (<1mg/mL at 25°C), DMF (sparingly), and PBS pH 7.2 (∼10 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 34369-07-8
Molecular Formula C10H14N5Na2O13P3
Molecular Weight (g/mol) 551.15
MDL Number MFCD00150755
InChI Key TTWYZDPBDWHJOR-WCYUCLFNNA-L
Synonym atp disodium salt hydrate, atp disodium salt, atp disodium hydrate, adenosine 5'-triphosphate disodium salt hydrate, atp disodium salt trihydrate, 56-65-5 non-sodium, 987-65-5 anhydrous, 5 inverted exclamation marka-atp-na2, adenosine 5'-triphosphate disodium sal
PubChem CID 131664345
IUPAC Name disodium;[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl] phosphate;hydrate
SMILES [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1

Especificaciones

Color White
Quantity 5 g
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,155
Solubility Information Soluble in water (110mg/ml at 25°C),ethanol (<1mg/ml at 25°C),DMSO (<1mg/ml at 25°C),DMF (sparingly),and PBS pH 7.2 (∽10mg/ml).
Formula Weight 551.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Adenosine-5'-triphosphate disodium salt hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM24L016 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberJ61125.09,J61125.06,J61125.14
Lot NumberX18J015 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberJ61125.09,J61125.06,J61125.14
Lot NumberM06J017 Certificate TypeCertificate of Analysis Date18/01/2023 Catalog NumberJ61125.09,J61125.06,J61125.14
Lot NumberZ01H039 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog NumberJ61125.09,J61125.06,J61125.14
Lot NumberN23I037 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog NumberJ61125.09,J61125.06,J61125.14

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Thermo Scientific Chemicals Adenosine-5'-triphosphate disodium salt hydrate, 98% >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado