missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenosine, 99+%

Product Code. 10071860
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-61-7
Molecular Formula C10H13N5O4
Molecular Weight (g/mol) 267.25
MDL Number MFCD00005752
InChI Key OIRDTQYFTABQOQ-KQYNXXCUSA-N
Synonym adenosine, adenocard, adenoscan, adenine riboside, adenosin, beta-d-adenosine, nucleocardyl, boniton, sandesin, myocol
PubChem CID 60961
ChEBI CHEBI:16335
IUPAC Name (2R,3R,4S,5R)-2-(6-aminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Absorbance 0.76 to 0.80 (A250/A260),0.14 to 0.16 (A280/A260)
Melting Point 233.0°C to 238.0°C
Color White
Loss on Drying 0.5% (105°C, 2 hrs) max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 26, V,16, 335
Merck Index 15141
Specific Rotation − 60.00
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
Spectrophotometry 98.0 to 102% (lambda max. 259nm,pH 7) molar absorptivity 15400+/-2% ,on dry basis
Transmittance 98% (c=0.5, H2O, 430nm) 1cm cell min.
Formula Weight 267.23
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Adenosine
Show More Show Less
Product Identifier
  • Adenosine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Adenosine, 99+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.