Learn More
3M™ Adflo™ Gas Filter A1B1E1 Kit
3M™ Adflo™ Gas Filter A1B1E1 Kit comes equipped with a heavy duty, lithium ion battery.
Brand: 3M™ 7000144540
225.00 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.97200kg Transport : UN number : 3481 Chem class : 9
Description
- A1B1E1 filter captures organic, inorganic and acid gases and vapors
- The heavy duty Lithium-ion battery fully charges in 5,5 hours and has a operating time of 12 hours
- When using the Adflo respirator with gas filter, you might require a breathing tube with silencer (83 40 08) and the heavy-duty battery (83 76 31)
Specifications
Turbo: approx. 10 to 13 hr. | |
Lithium Ion | |
A1B1E1 | |
Heavy duty |