Learn More
3M™ Adflo™ Gas Filter A2
3M™ Adflo™ Gas Filter A2 is designed to capture organic, inorganic, and acid gases and vapors.
Brand: 3M™ 7000028075
77.03 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- When using the Adflo respirator with gas filter, you might require a breathing tube with silencer (83 40 08) and the heavy-duty battery (83 76 31)
- Replace the gas filter according to your work place requirements
- It is not recommended to wait until you can feel odors and smell
Specifications
Organic Vapor, Acid Gas | |
Black | |
Gas Filter |