missing translation for 'onlineSavingsMsg'
Learn More

Color Coded High Resolution pH Buffer, Reagecon™

Use color coded pH buffers for ease of use. Reagecon™ Color Coded High Resolution pH Buffers are tested at either 20°C or 25°C and are certified and NIST traceable.

Brand:  Reagecon H40525C

Code : NEW

Additional Details : Weight : 0.51570kg

 View more versions of this product

Product Code. 11494599

  • £33.60 / 500mL
Estimated Shipment: 15-08-2024
to see stock.

Description

Description

  • Colored pH Buffer Solutions
  • High Resolution
  • Tested at 20°C or 25°C 
  • Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
  • NIST traceable
  • Presented in various pack sizes
Specifications

Specifications

0°C
4
500 mL
Red
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers