Learn More
ITW Chemische Produkte™ CleanStep™ Adhesive Cleanroom Mats
Reduce contamination in high traffic areas. ITW Chemische CleanStep™ Adhesive Cleanroom Mats are constructed with either 30 or 60 layers. Each removable sheet has a consistent adhesive coating designed to trap particles and prevent transfer.
Brand:  ITW Chemische Produkte™ AMA184681G
 View more versions of this product
Product Code. 11595315
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Designed to capture particulates from foot and wheel borne traffic prior to entry into cleanrooms or controlled environments
  • Provide a portable contamination control solution
  • Can be used with or without frames (sold separately)
  • Contain anti-microbial preservatives that prevent microbial growth
  • Consistent adhesive coating on each layer of 30 and 60 layer mats
  • Possible applications include cleanroom and critical environment entry points, gowning rooms or anterooms in the semiconductor, biotechnology, medical device, pharmaceutical processing, aerospace, and automotive markets
Specifications
Specifications
18 x 46 in. (46 x 117 cm)
30 layer
Gray
Polyethylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
ITW Chemische Produkte™ CleanStep™ Adhesive Cleanroom Mats > Size: 18 x 46 in. (46 x 117cm); 30 layers; Gray