missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Adhesive Labels
Transparent, punched labels for use with wash bottles and plastic or glass reagent bottles
Brand: Kartell™ 0501800
65.67 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with wash bottles and plastic or glass reagent bottles, as recommended by the NFPA (National Fire Protection Agency)
- Complete with risk assessment numbers, recommendations for use and special notices shown in reagent catalogues
Specifications
Transparent with colored text | |
Methylene Cloride | |
130 x 35 mm |
Black | |
50/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction