missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Adhesive Labels
Transparent, punched labels for use with wash bottles and plastic or glass reagent bottles
Brand: Kartell™ 0501000
68.12 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Additional Details : Weight : 7.65000kg
Description
- For use with wash bottles and plastic or glass reagent bottles, as recommended by the NFPA (National Fire Protection Agency)
- Complete with risk assessment numbers, recommendations for use and special notices shown in reagent catalogues
Specifications
Transparent with colored text | |
Dist. Water | |
130 x 35 mm |
Blue | |
50/Pk. |