missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Adhesive Labels

Transparent, punched labels for use with wash bottles and plastic or glass reagent bottles

Brand:  Kartell™ 0501200

67.88 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 15855517

  • £74.70 / Pack of 50
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with wash bottles and plastic or glass reagent bottles, as recommended by the NFPA (National Fire Protection Agency)
  • Complete with risk assessment numbers, recommendations for use and special notices shown in reagent catalogues
Specifications

Specifications

Transparent with colored text
Methanol
130 x 35 mm
Black
50/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers