Learn More
Thermo Scientific™ ABgene™ Adhesive PCR foil seal
Brand: Thermo Scientific™ ABgene™ AB-0626-RMP340
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Strong adhesive prevents evaporation during thermal cycling or sample storage
- Sample is accessed by piercing foil with a pipettor tip
- No adhesive build-up on tip after piercing
- End tabs for easy seal application and removal
- Recommended for use in conjunction with a thermal cycler that has a screw or clip down lid
- Temperature range: -80°C to 150°C