missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adipic dihydrazide, 98%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | 178.0°C to 182.0°C |
| Color | White to Yellow |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Plastic bottle |
| Linear Formula | H2NNHCO(CH2)4CONHNH2 |
| Quantity | 100 g |
| Beilstein | 02, I, 277 |
| Show More |
Safety and Handling
Product Identifier
- Adipic dihydrazide
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Respiratory sensitiser Category 1
- H411-Toxic to aquatic life with long lasting effects.
- H317-May cause an allergic skin reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction