missing translation for 'onlineSavingsMsg'
Learn More
Adiponitrile, 98%, Thermo Scientific Chemicals Product Code.: 11399799

Adiponitrile, 98%, Thermo Scientific Chemicals

Product Code. 11399799
1000 mL
Click to view available options
Quantity:
250 mL
1000 mL
This item is not returnable. View return policy

Product Code. 11399799

Brand: Thermo Scientific Alfa Aesar B23122.AU

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-69-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00001975
InChI Key BTGRAWJCKBQKAO-UHFFFAOYSA-N
Synonym adiponitrile, 1,4-dicyanobutane, adipodinitrile, adipyldinitrile, adipic acid dinitrile, tetramethylene dicyanide, nitrile adipico, tetramethylene cyanide, adipic acid nitrile, adipinsaeuredinitrilShow MoreShow Less
PubChem CID 8128
IUPAC Name hexanedinitrile
SMILES C(CCC#N)CC#N

Specifications

Melting Point 2°C
Density 0.951
Boiling Point 295°C
Flash Point 163°C (325°F)
Odor Odorless
Refractive Index 1.438
Quantity 1000 mL
UN Number UN2205
Beilstein 1740005
Formula Weight 108.14
Percent Purity 98%
Chemical Name or Material Adiponitrile
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252544 Certificate TypeCertificate of Analysis Date23/09/2024 Catalog NumberB23122.AU,B23122.AK
Lot NumberFA010187 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23122.AU,B23122.AK
Lot Number18193 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23122.AU,B23122.AK
Lot NumberH16X002 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23122.AU,B23122.AK
compliance-icons
Product Identifier
  • Adiponitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
Hazard Statement
  • H301-Toxic if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : Hexanedinitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Adiponitrile, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.