missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adipoyl chloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Adipoyl chloride is used in the production of nylon by reacting with hexamethylene diamine. It is also used in the preparation of biphenyl end-capped liquid crystals. Further, it is employed in the synthesis of chiral polymer for membrane applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Adipoyl chloride is used in the production of nylon by reacting with hexamethylene diamine. It is also used in the preparation of biphenyl end-capped liquid crystals. Further, it is employed in the synthesis of chiral polymer for membrane applications.
Solubility
Immiscible with water.
Notes
Incompatible strong oxidizing agents, bases and acids.
Adipoyl chloride is used in the production of nylon by reacting with hexamethylene diamine. It is also used in the preparation of biphenyl end-capped liquid crystals. Further, it is employed in the synthesis of chiral polymer for membrane applications.
Solubility
Immiscible with water.
Notes
Incompatible strong oxidizing agents, bases and acids.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.255 |
Boiling Point | 105°C to 107°C (2 mmHg) |
Flash Point | >110°C (230°F) |
Odor | Acrid |
Linear Formula | ClCO(CH2)4COCl |
Refractive Index | 1.4705 |
Quantity | 100 g |
UN Number | UN3265 |
Beilstein | 507709 |
Sensitivity | Moisture sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'