Learn More
Siemens ADIVA™ Sheath Rinse
Universal rinse for use with the ADIVA 120 / 2120 / 2120i Hematology Systems. Siemens ADIVA™ Sheath Rinse is available in a 20L container, sufficient for 1400 tests.
Brand: Siemens T01-3623-01
Code : Z1
Additional Details : Weight : 120.00000kg
Description
The ADVIA™ 120 System offers a unique portfolio of diagnostic assays and gold-standard flow cytometry testing. To satisfy the demands of laboratories, Siemens delivers a hematology analyzer with comprehensive testing parameters, flexible sample handling, a user-defined interface, and a compact bench-top design.
- Automates hematology workflow without the need for large track-based systems or reflexive testing
- Maximizes efficiency through user-defined screen configurations and reports
- Simplifies maintenance with Unifluidics™ Technology through reduced fluidics, elimination of pinch valves, and automated daily cleaning
- Streamlines productivity with a 150 capacity rack-based autosampler, allowing 75 minutes of true walk-away time
- Reduces the need for manual slide production and review by utilizing WBC and RBC/PLT technology and high specificity
- Maximizes the effectiveness of costly platelet transfusions with accurate results the first time—even at very low platelet levels
Specifications
Colorless | |
ADIVA™ 120 / 2120 / 2120i Hematology Systems | |
7.3 | |
Liquid |
Universal rinse | |
Shealth Rinse | |
20 L |