missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ QuantiGene™ Plex Human ADMETOX Panel, 80-plex

The QuantiGene Plex Human ADMETOX Panel, 80-plex, enables the measurement of gene expression of all relevant ADME (absorption, distribution, metabolism, and excretion) and toxicological biomarkers through the analysis of 80 RNA targets in a single well using Luminex xMAP technology in combination with branched DNA signal amplification.

Brand:  Invitrogen™ QGP-280-ADMETOX

 View more versions of this product

Product Code. 17912011

  • £20866.00 / Pack of 3

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The QuantiGene Plex Human ADMETOX Panel, 80-plex enables the measurement of gene expression of all relevant ADME (absorption, distribution, metabolism, and excretion) and toxicological biomarkers through the analysis of 80 RNA targets in a single well using Luminex xMAP technology in combination with branched DNA signal amplification. Analyze 80 genetic targets implicated in drug metabolism, including enzymes, transporters, and CYPs, as well as toxicology and apoptosis for efficient profiling of your drug discovery and basic research samples.

QuantiGene Plex panels are extensively tested for target combinability, interference, and cross-reactivity to provide the highest level of validation and precision. This kit is not compatible with the MAGPIX instrument.

This panel is used in combination with the QuantiGene Plex Assay Kit (Cat. No. QP1013), which contains all buffers, plates, and detection reagents necessary to perform the assay in 96-well plates.

Features of the QuantiGene Plex Human ADMETOX Panel, 80-plex include:

  • True multiplexing—measure 80 genes of interest and housekeeping genes in the same well with no cross-reactivity
  • Accommodation of difficult sample types—works with degraded and cross-linked RNA in FFPE tissues and blood
  • Standardized platform—uses 96-well plate format compatible with Luminex 200, FLEXMAP 3D, and INTELLIFLEX systems (384-well format available on demand)
  • Simple workflow—employs an ELISA-like workflow with direct hybridization of transcripts to beads and transcript labeling (branched DNA signal amplification and PE readout)
  • Customize with validated targets—if alternative targets are needed in this panel, select from our large inventory of over 22,000 genes to create custom research panels
  • Rapid turnaround for this catalog panel for fast delivery

Targets (80-plex): ABCA1, ABCB1 , ABCB11, ABCC1, ABCC2, ABCC3 , ABCG2 , ACTB, AOX1, B2M, BAX, BCL2, CASP3,CASP9, CES1, CYP1A1, CYP1A2, CYP2A6, CYP2B6, CYP2C19, CYP2C8, CYP2C9, CYP2D6, CYP2E1, CYP2J2, CYP3A4, CYP3A5, CYP3A7, DPYD, FMO1, FMO3, FMO5, GAPDH, GSTA2, GSTM1, GSTP1, GSTT1, GUSB, HMGB1, HMGCS2, HNF4A, HPRT1, NAT1, NAT2, PGK1, POLR2A, PPIB, RIPK1, RXRA, SLC10A1, SLC15A1, SLC15A2, SLC16A1, SLC22A1, SLC22A2, SLC22A6, SLC22A7, SLC2A1, SLC3A1, SLCO1B1, SLCO1B3, SLCO2B1, SULT1A1, SULT2A1, SULT2B1, TBP, TFRC, TP53, TPMT, UGT1A1, UGT1A10, UGT1A3, UGT1A4, UGT1A6, UGT1A9, UGT2B15, UGT2B17, UGT2B4, UGT2B7, XDH

Panel and signal amplification technology

QuantiGene Plex assays are hybridization-based assays for the direct quantitation of RNA from cell lysates, tissue homogenates, or purified RNA. They enable the measurement of multiple targets in the same well via multiplexing.

QuantiGene Plex panels contain two components: a target-specific probe set and magnetic capture beads. The target-specific probes are used to capture the RNA of interest, and branched DNA technology then amplifies the fluorescent signals, which are read with a Luminex xMAP instrument. The fluorescent signal is directly proportional to the amount of target mRNA present in the sample. This allows for (relative) quantification of gene expression and with this specific panel enables human immune profiling. Detailed and step-by-step data analysis is facilitated by the QuantiGene Plex data analysis app.

Applications

QuantiGene Plex assay research applications include compound screening of drug candidates, biomarker verification, siRNA knockdown efficiency, prospective and/or retrospective analysis of clinical samples, microarray validation, predictive toxicology, and detection of translocations and fusion genes. In general, any multiplex gene expression analysis can be done with QuantiGene Plex assays, independent of species or sample type.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

96-well
Luminex Instrument
QuantiGene™ Plex
3 Plates
Human
Fluorescence
QuantiGene Gene Expression Assay

Target-specific Probe Set and magnetic Capture Beads (premixed and ready to use).

Store Probe Set at -20°C and Capture Beads at 4°C (do not freeze).

Multiplex Panel
Wet Ice
Purified RNA, cell/blood/FFPE lysates, tissue homogenates
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.