missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adonitol, 98%

Adonitol, 98%, C5H12O5, CAS Number-488-81-3, adonit, adonite, adonitol, d-ribitol, d-xylitol, eutrit, ribitol, xylit, xylite, xylitol, 100g, 97.5% min. (ELSD) (HPLC), 01,53, 100.0, 99.0, White, 207-685-7, 152.15, HEBKCHPVOIAQTA-NGQZWQHPSA-N, Authentic, (2S,4R)-pentane-1,2,3,4,5-pentol

£32.10 - £879.00

Chemical Identifiers

CAS 488-81-3
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064291
InChI Key HEBKCHPVOIAQTA-NGQZWQHPSA-N
Synonym xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
PubChem CID 6912
IUPAC Name (2S,4R)-pentane-1,2,3,4,5-pentol
SMILES C(C(C(C(CO)O)O)O)O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10657332
View Documents
Thermo Scientific Chemicals
167030050
5 g Glass bottle
£32.10
5g
Please to purchase this item. Need a web account? Register with us today!
10667332
View Documents
Thermo Scientific Chemicals
167030100
10 g Glass bottle
£54.40
10g
Please to purchase this item. Need a web account? Register with us today!
10793085
View Documents
Thermo Scientific Chemicals
167031000
100 g Glass bottle
£177.00
100g
Please to purchase this item. Need a web account? Register with us today!
11904641
View Documents
Thermo Scientific Chemicals
167035000
500 g Glass bottle
£879.00
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

488-81-3
152.15
HEBKCHPVOIAQTA-NGQZWQHPSA-N
6912
C(C(C(C(CO)O)O)O)O
C5H12O5
MFCD00064291
xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
(2S,4R)-pentane-1,2,3,4,5-pentol

Specifications

488-81-3
100.0
White
97.5% min. (ELSD) (HPLC)
MFCD00064291
15,157
Solubility in water: soluble. Other solubilities: soluble in hot alcohol,insoluble in ether
C(C(C(C(CO)O)O)O)O
152.15
152.15
Fine Crystalline Powder
99.0
101.0°C to 104.0°C
Authentic
C5H12O5
01,53
xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
HEBKCHPVOIAQTA-NGQZWQHPSA-N
(2S,4R)-pentane-1,2,3,4,5-pentol
6912
98%
Adonitol
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 207-685-7

Product Identifier
  • Adonitol

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only