missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adonitol, 99.8%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Color | White |
| Density | 1.53g/cm3 |
| Assay Percent Range | 0.998 |
| Quantity | 100 g |
| Formula Weight | 152.15 |
| Physical Form | Powder |
| Chemical Name or Material | Adonitol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Adonitol, 99.8%, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction
