missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clinitrol™ 290 Reference Solution; Advanced Instruments

Critical normal human serum midpoint of 290mOsm
£130.38 - £192.00
Specifications
Certifications/Compliance | CE |
---|---|
Color | Clear |
Content And Storage | 2°C to 30°C |
Concentration | 290 mOsm/kg H2O |
Sample Type | Serum |
Product Code | Brand | For Use With (Equipment) | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Quantity | Price | Quantity & Availability | |||||
13214750
![]() |
N/A
3LA029 |
3D3, 3250 and 3900 osmometers | 10 x 5 mL | |||||||
12802201
![]() |
N/A
3MA029 |
2020, 3300 and 3320 Osmometers | 10 x 2 mL | |||||||
13112093
![]() |
N/A
3MA029 |
2020, 3300 and 3320 Osmometers | 10 x 2 mL | |||||||
Description
Critical normal human serum midpoint of 290mOsm
- Checks performance of freezing-point osmometers in the critical area of normal serum values
- Contains known and controlled quantities of some of the same solutes which comprise osmotically active particles of serum
- Pipetted into a tube and run just like normal serum
- Runs as the first and last samples in a series of tests to monitor operator technique and instrument performance
- Does not require mixing, reconstituting or refrigerating; one-year shelf life
Specifications
CE | |
2°C to 30°C | |
Serum | |
Sodium Chloride (NaCl) and 2-Hydroxybiphenyl Sodium Salt Tetrahydrate | |
Liquid |
Clear | |
290 mOsm/kg H2O | |
Reference Solution | |
Osmometer instrument calibration and quality control |
Safety and Handling
missing translation for 'shelfLife' : 3 Years
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Clinitrol™ 290 Reference Solution; Advanced Instruments