Learn More
Advanced Instruments Osmometer Calibration Standards
Calibration standards for use with Advanced osmometers
Brand:  Advanced Instruments 3LA051
Encompass
Product Code. 12822733
  • £160.50 / Pack of 10
Description
Calibration standards for use with Advanced osmometers
  • Range from 50 to 3000 mOsm/kg
Specifications
Specifications
Clear
Sodium Chloride 0.15% to 9.5%, 2-Hydroxybiphenyl Sodium Salt Tetrahydrate <0.1%
10 x 5 mL
Room Temperature
Osmometer instrument calibration and quality control