Learn More
Advanced Instruments Probe Wiper Rings
Probe Wiper Rings for Advanced Multi-Sample Osmometer, Model 2020
Brand:  Advanced Instruments 202840
Encompass
Product Code. 13182083
  • £261.00 / Pack of 50
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with Advanced osmometers
Specifications
Specifications
Probe Wiper Ring
Advanced Model 2020 and Fiske Model 2400