missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Advantec 301900 Pressure Filtration Holder

Process samples from an in-line system or pressure vessel with this stainless steel holder. Cole-Parmer™ Advantec 301900 Pressure Filtration Holders maximize flow rates by applying positive pressure to the fluid over a broad membrane surface.

Brand:  Cole-Parmer™ 02927-50

Product Code. 11716575

  • £4780.00 / Each
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 304 Stainless Steel Pressure Filtration Holder for 142mm filter 
  • Holders are autoclavable with one filter membrane that is sealed with removable wing nuts 
  • All holders vent to release pressure
  • Holders have PTFE gaskets, silicone O-rings, and PTFE-coated photoetched stainless steel support screens (model 02926-50 has a 304 SS support screen) to prevent nitrocellulose membranes from sticking
Specifications

Specifications

3/4 in NPT to 11mm ID (adapter included)
Stainless Steel (304)
100psi (7 bar)
42.55 cm
142mm filter
113 cm2
16.75 in.
Filter: 142mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Advantec 301900 Pressure Filtration Holder > Cole-Parmer Advantec 301900 Pressure Filtration Holder

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.