Learn More
Cole-Parmer™ Advantec™ GC50 Borosilicate Glass Fiber Filter
Use as pre-filters to extend membrane life, for water/air pollution analysis or liquid clarification. Advantec™ GC50 Borosilicate Glass Fiber Filters are ideal for analytical or gravimetric studies.
Brand:  Cole-Parmer™ 06656-03
Additional Details : Weight : 0.06300kg
Product Code. 12726298
  • £128.00 / Pack of 100
In Stock
to see stock.
Add to basket
Description
Description
Borosilicate glass filters create a biologically inert filter matrix that is resistant to both chemicals and pH extremes. Filters without binder withstand up to 500°C. Filters with binder withstand up to 120°C. Grades GC90, GS25 and DP70 with binder offer higher wet strength and lower fiber release.
Specifications
Specifications
Borosilicate Glass Filter
0.5 μm
100 Pack
75 mm
Borosilicate Glass
For Water/air Pollution Analysis, or Liquid Clarification
Filter