missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Adventurer™ Precision Balance

Product Code. 15325063 Shop All Ohaus Products
Click to view available options
Capacity:
220 g
420 g
520 g
620 g
820 g
1500 g
2200 g
4200 g
5.2 kg
6200 g
8200 g
12 kg
Readability:
0.001 g
1 mg
0.01 g
0.1 g
Model:
AX12001
AX12001/E
AX12001M
AX1502
AX1502/E
AX1502M
AX2201
AX2201/E
AX2202
AX2202/E
AX2202M
AX223
AX223/E
AX223M
AX4201/E
AX4202
AX4202/E
AX4202M
AX422/E
AX423
AX423/E
AX423M
AX5202
AX5202M
AX523
AX523/E
AX523M
AX6202
AX6202/E
AX6202M
AX623
AX623/E
AX623M
AX8201/E
AX8201M
AX822/E
Calibration:
External
Internal
missing translation for 'unitSize'
1 unit
Each
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 15325063

missing translation for 'mfr': OHAUS™ 30127969

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Intuitive Adventurer™ Precision Balance designed for routine weighing.

Intuitive Balance Designed for Routine Weighing:

Striking the ideal balance between inventive features and functional, uncomplicated weighing capabilities, the OHAUS Adventurer incorporates all of the applications necessary for routine weighing and measurement activities. With a color touchscreen, three level user management to fulfill GLP/GMP compliance capabilities, two USB ports, and much more, Adventurer is the most complete balance in its class.

Unique Features Include:

  • Adventurer balances feature a color touchscreen, icon-based user interface, and an ergonomic design -making them easy to configure and use.
  • Features such as specialized weighing modes, multiple connectivity options, and AutoCal™ provide versatility and flexibility for a variety of applications.
  • Durable construction, large weighing surfaces, a space-saving draftshield design, and full housing in-use cover allow for use in lab, education and industrial environments.

Stability, Accuracy, and Fast Operation Ensure Optimal Weighing Results in Routine Weighing Tasks:

Weighing Performance:

  • Delivers stable and reliable weighing results for routine weighing tasks.

Stabilization Time:

  • The fast stabilization time of the Adventurer improves productivity in the laboratory.

Calibration:

  • AutoCal™ — Selected models feature the OHAUS automatic internal calibration system that performs routine maintenance by calibrating the balance daily.
  • External Calibration — Traditional calibration in which the operator manually calibrates the balance with their choice of calibration weight value to ensure accuracy available on every model.

Color Touchscreen Offers Easy and Fast Operation of Adventurer Applications

  • Operate and access nine Adventurer application modes and abundant features that eliminate the need to do several manual calculations through the modern color touchscreen.
  • Operators can wear laboratory gloves while utilizing the touchscreen, eliminating the inconvenience and hazards associated with constantly putting on and removing gloves.
  • In addition to the touchscreen, Adventurer also has six mechanical keys that provide tactile feedback and allow the operator to perform repetitive operations such as tare, zero, calibration, and print.

Equipped with the Connectivity and Functional Features Required in Laboratories

Dual USB Ports:

  • A front USB host port is easily accessible and makes it simple to load data from the balance on to a flash drive without having to reach around to the back or move the balance.
  • A second USB device is located at the rear of the balance that can be used to connect the balance to a PC.
  • The connectivity options help meet traceability requirements in traditional installations.

Label Printing Function:

  • Easy to link with Zebra printer and have one built-in label printing template.

Balance Profiles:

  • The cloning feature allows you to save user and application settings to a USB flash drive which can be easily used to configure additional Adventurer balances.

Below Minimum Sample Weight Indication:

  • When using the minimum weight feature, the display clearly indicates that your current sample weight is below your defined minimum limit. Simply increase your sample weight to assure that your results are up to your standard.

Space-saving Draftshield Designed to Improve User Experience and Accessibility:

  • Draftshield doors are constructed of two glass panels, reducing the space required on the lab bench when the doors are open.
  • Wide door entry provides unobstructed access and allows larger weighing vessels to be easily placed on the pan, reducing the chance of accidental spillage.
  • Easy to keep clean in order to ensure a safe workspace by minimizing contamination.

Power Saving Functions:

  • The Adventurer is designed with various power-saving features that help reduce the environmental impact.
  • These include auto-off and brightness controls.

User Management Function:

  • Ensures data security and data traceability.
  • 3 level user management function ensures data security and data traceability requirements.
  • One administrator, two supervisors and 10 users have preset accessibility in the software.

Real Time Clock with GLP/GMP Data:

  • A real-time clock function keeps accurate time even during power loss.
  • GLP data capability has the ability to record Sample name, Project names and Balance IDs to help meet traceability and compliance requirements.
TRUSTED_SUSTAINABILITY

Spezifikation

Certifications/Compliance Metrology: OIML R76, EN 45501 (Class I, nmax 320000, Class II, nmax 52000), Product Safety: EN 61010-1, IEC 61010-1, Electromagnetic Compatibility: IEC 61326-1, EN 61326-1 (Emissions Class B, Immunity Industrial Requirements), GLP/GMP Compliant
Connection Type RS232, USB Host, USB Device
Description Precision Balance AX2202/E
Capacity 2200 g
Display Type Color Touchscreen
Readability 0.01 g
For Use With (Application) Weighing, Parts Counting, Percent Weighing, Check
Includes AC Adapter
Linearity ±0.02 g
Material Metal Base, ABS Top Housing, Stainless Steel Pan
Measurement Time 1.5 sec.
Min. Sample Weight 8.2 g
Model AX2202/E
Repeatability 0.01 g
Sensitivity Drift 3 ppm/°C
Stabilization Time 1.5 sec.
Temperature Range (English) 50°F to 86°F
Temperature Range (Metric) 10°C to 30°C
Test Load 2000 g, OIML F1 or ASTM CL 2
Type Precision Balance
Warranty 2 years
Weighing Modes Weighing (17 units + custom unit), parts counting, percentage weighing, check weighing, animal/dynamic weighing, totalization/statistics, formulation, density determination, display hold, batch printing
Dimensions (L x W x H) 354 x 230 x 100 mm
Length (English) 13.93 in.
Length (Metric) 354 mm
Width (English) Pan 6.9 in.
Width (Metric) 230 mm
Width (Metric) Pan 175 mm
Height (Metric) 100 mm
Depth (English) Pan 7.7 in.
Depth (Metric) 354 mm
Depth (Metric) Pan 195 mm
Weight (English) 13.2 lb.
Weight (Metric) 6 kg
Electrical Requirements AC Adapter Input: 100/240 V, 0.3 A, 50/60 Hz
Voltage 100/240 V
Amperage AC Adapter Input: 0.3A, AC Adapter Output: 0.84A
Frequency AC Adapter Input: 50/60 Hz
Legal for Trade No
Calibration External
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt