Learn More
OHAUS™ Adventurer™ Precision Balance
Intuitive Adventurer™ Precision Balance designed for routine weighing.
Brand:  OHAUS™ 30805890
2545.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17941292
  • £2640.00 / 1 unit
Explore more special offers
Description
Show More
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
OHAUS™ Adventurer™ Precision Balance > Model: AX623, Capacity: 620.0 g, Readbility: 0.001 g, Calibration: Internal