Learn More
Axygen™ Aerosol Filter Tips, Sterile
Prevent cross-contamination with robotic barrier tips. Axygen™ Aerosol Filter Tips, Sterile are e-beam irradiated to prevent product degradation and are free of leachable additives. Available in a wide variety of tip styles.
£97.40 - £870.00
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Color | Clear |
Clean Claims | DNase, RNase, and Pyrogen Free |
Material | Polypropylene |
Product Code | Brand | Volume (Metric) | Sterility | Tip Style | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Sterility | Tip Style | Price | Quantity & Availability | |||
12746709
|
Axygen™
MTXF-1250-C-R-S |
15 to 1250 μL | Sterile | Matrix-style |
£600.00
Pack of 3456 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
12571348
|
Axygen™
TF-100 |
1 to 100 μL | Sterile | Universal |
£870.00
Pack of 10000 |
In Stock
Log in to see stock. |
|||
12673706
|
Axygen™
TF-1000-L-R-S |
100 to 1000 μL | Sterile | Maxymum Recovery |
£161.00
Pack of 1000 |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||
11356084
|
Axygen™
TF-100-R-S |
1 to 100 μL | Sterile | Universal |
£119.00
Pack of 960 |
In Stock
Log in to see stock. |
|||
11306144
|
Axygen™
TF-1000-R-S |
100 to 1000 μL | Sterile | Universal |
£155.00
Pack of 1000 |
In Stock
Log in to see stock. |
|||
12685876
|
Axygen™
TF-100-L-R-S |
1 to 100 μL | Sterile | Maxymum Recovery |
£177.00
Pack of 960 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
11336144
|
Axygen™
TF-1200-C-R-S |
100 to 1100 μL | Sterile | Universal |
£151.00
Pack of 1152 |
In Stock
Log in to see stock. |
|||
12689595
|
Axygen™
TF-200-L-R-S |
1 to 200 μL | Sterile | Maxymum Recovery |
£161.00
Pack of 960 |
In Stock
Log in to see stock. |
|||
11326064
|
Axygen™
TF-200-R-S |
1 to 200 μL | Sterile | Universal |
£148.00
Pack of 960 |
In Stock
Log in to see stock. |
|||
12663826
|
Axygen™
TF-20-L-R-S |
0.5 to 20 μL | Sterile | Maxymum Recovery |
£161.00
Pack of 960 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
11341904
|
Axygen™
TF-20-R-S |
0.5 to 20 μL | Sterile | Universal |
£115.00
Pack of 960 |
In Stock
Log in to see stock. |
|||
12641656
|
Axygen™
TF-350-L-R-S |
1 to 300 μL | Sterile | Maxymum Recovery |
£106.00
Pack of 960 |
In Stock
Log in to see stock. |
|||
11376074
|
Axygen™
TF-350-R-S |
1 to 300 μL | Sterile | Universal |
£122.00
Pack of 960 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
11304285
|
Axygen™
TF-420-L-R-S |
0.5 to 20 μL | Sterile | Maxymum Recovery, Ultra Micro |
£112.00
Pack of 960 |
In Stock
Log in to see stock. |
|||
10068559
|
Axygen™
TF-420-R-S |
0.5 to 20 μL | Sterile | Ultra-micro |
£470.00
Pack of 4800 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||
12675876
|
Axygen™
TF-50-L-R-S |
0.5 to 50 μL | Sterile | Maxymum Recovery |
£111.00
Pack of 960 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
12673826
|
Axygen™
TF-50-R-S |
0.5 to 50 μL | Sterile | Universal |
£97.40
Pack of 960 |
In Stock
Log in to see stock. |
|||
12746739
|
Axygen™
TF-200-R-S |
1 to 200 μL | Sterile | Universal |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12746729
|
Axygen™
TF-1000-R-S |
100 to 1000 μL | Sterile | Universal |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12776739
|
Axygen™
TF-20-L-R-S |
0.5 to 20 μL | Sterile | Maxymum Recovery |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12575667
|
Axygen™
TF-1200-C-L-R-S |
100 to 1100 μL | Sterile | Maxymum Recovery |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12796729
|
Axygen™
TF-1200-C-L-R-S |
100 to 1100 μL | Sterile | Maxymum Recovery |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12786739
|
Axygen™
TF-20-R-S |
0.5 to 20 μL | Sterile | Universal |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12786729
|
Axygen™
TF-100-R-S |
1 to 100 μL | Sterile | Universal |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12716749
|
Axygen™
TF-350-L-R-S |
1 to 300 μL | Sterile | Maxymum Recovery |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12726749
|
Axygen™
TF-350-R-S |
1 to 300 μL | Sterile | Universal |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12776729
|
Axygen™
TF-100-L-R-S |
1 to 100 μL | Sterile | Maxymum Recovery |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12706739
|
Axygen™
TF-1200-C-R-S |
100 to 1100 μL | Sterile | Universal |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12776749
|
Axygen™
TF-420-L-R-S |
0.5 to 20 μL | Sterile | Maxymum Recovery, Ultra Micro |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11350105
|
Axygen™
MTXF-1250-C-R-S |
15 to 1250 μL | Sterile | Matrix-style |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12716759
|
Axygen™
TF-50-L-R-S |
0.5 to 50 μL | Sterile | Maxymum Recovery |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12736739
|
Axygen™
TF-200-L-R-S |
1 to 200 μL | Sterile | Maxymum Recovery |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12726759
|
Axygen™
TF-50-R-S |
0.5 to 50 μL | Sterile | Universal |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12736729
|
Axygen™
TF-1000-L-R-S |
100 to 1000 μL | Sterile | Maxymum Recovery |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Technologically advanced filter barrier pipette tips
- Ideal for DNA amplification and microbiological applications where trace contamination can invalidate results
- Filter allows free passage of air while preventing cross-contamination by blocking the passage of aerosols or liquids
- Nonpyrogenic, clear polyethylene filters are free from DNA inhibitors and leachable additives
- Every filter is inserted into the tip robotically to ensure freedom from nuclease contamination
- Packaged in sealed-bottom racks with lids to prevent contamination
- Wide variety of tip styles available in a range of packaging quantities:
- Maxymum Recovery™ technology provides excellent accuracy and prevents wastage of precious samples
- Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
Order Info
Available in bulk or racked formats.
Certifications
Certified RNase-, DNase- and Pyrogen free
Specifications
Filtered | |
Clear | |
Polypropylene |
Sterile | |
DNase, RNase, and Pyrogen Free |