missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Aerosol-Tight Caps
Use this replacement lid with 100mL round buckets. Eppendorf™ Replacement Lids are aerosol- tight.
Brand: Eppendorf™ 5702721000
60.32 GBP valid until 2024-11-30
Use promo code "21681" to get your promotional price.
This item is not returnable.
View return policy
Description
- Set of 2 caps
- Aerosol tight
- For 100mL round buckets
Specifications
Cap | |
100mL Round Bucket |