Learn More
Thermo Scientific™ Aerosol-Tight Lid
Aerosol-Tight Lid for 75007570 Round Bucket, for use with Rotor S-4-104, Rotor S-4x750 and Rotor S-4x1000.ids
Brand: Thermo Scientific™ 75007584
Code : 51
Additional Details : Weight : 2.50000kg
Description
Aerosol-Tight Lid for 75007570 Round Bucket, for use with Rotor S-4-104, Rotor S-4x750 and Rotor S-4x1000.ids
- Aerosol-Tight Lid
- For 75007570 Round Bucket
- For use with Rotor S-4-104, Rotor S-4x750 and Rotor S-4x1000.ids
Specifications
Aerosol-tight Lid | |
Rotor S-4-104, Rotor S-4 x 750 and Rotor S-4 x 1000.Ids, 75007570 Round Bucket | |
750 mL bucket in S-4-104 rotor with 5810/5810 R centrifuge |