missing translation for 'onlineSavingsMsg'
Learn More
AF-DX 116, Tocris Bioscience™ Product Code.: 18782684

AF-DX 116, Tocris Bioscience™

Product Code. 18782684
50 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18782684

Brand: Tocris Bioscience 1105/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective M2 antagonist

Selective M2 muscarinic receptor antagonist. Ki values are 64, 417, 786, 211 and 5130 nM for human recombinant M2, M1, M3, M4 and M5 muscarinic receptors, respectively.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102394-31-0
Molecular Formula C24H31N5O2
Molecular Weight (g/mol) 421.545
InChI Key UBRKDAVQCKZSPO-UHFFFAOYSA-N
Synonym otenzepad, af-dx 116, af-dx-116, unii-sx2wtg9r3z, unii-ww6a9tfl2c, sx2wtg9r3z, ww6a9tfl2c, chembl17045, 11-2-diethylamino methyl-1-piperidinyl acetyl-5,11-dihydro-6h-pyrido 2,3-b 1,4 benzodiazepin-6-one, 11-2-2-diethylamino methyl piperidin-1-yl acetyl-5h-benzo e pyrido 3,2-b 1,4 diazepin-6 11h-oneShow MoreShow Less
PubChem CID 107867
IUPAC Name 11-[2-[2-(diethylaminomethyl)piperidin-1-yl]acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one
SMILES CCN(CC)CC1CCCCN1CC(=O)N2C3=CC=CC=C3C(=O)NC4=C2N=CC=C4

Specifications

Quantity 50 mg
Formula Weight 421.54
Percent Purity >98%
Chemical Name or Material AF-DX 116
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
AF-DX 116, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.