missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AF-DX 384, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent M2/M4 selective antagonist (pKi values are 8.22, 8.00, 7.51, 7.18 and 6.27 at human M2, M4, M1, M3 and M5 receptors respectively).
Specifications
Quantity | 10 mg |
Formula Weight | 478.64 |
Percent Purity | >98% |
Chemical Name or Material | AF-DX 384 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'