Learn More
AffiniPure Goat anti-Rabbit, Biotin, Polyclonal, Proteintech
Goat Polyclonal Antibody
Brand: Proteintech SA00004-2-200UL
This item is not returnable.
View return policy
Description
AffiniPure Polyclonal antibody specifically detects AffiniPure in Human, Mouse, Rat samples. It is validated for ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
AffiniPure | |
Polyclonal | |
Biotin | |
200 μL | |
Human, Mouse, Rat | |
IgG |
ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry | |
0.3 mg/mL | |
Goat | |
Secondary | |
Liquid |