missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ ExoSAP-IT™ For PCR Product Clean-Up
Click to view available options
Quantity:
10 mL
4 x 1 mL
Unit Size:
2000 reactions
5000 reactions
Description
- Two hydrolytic enzymes, Exonuclease I and Shrimp Alkaline Phosphatase (SAP), together in a specially formulated buffer
- 30-minute protocol
Specifications
Specifications
| Content And Storage | -20°C |
| For Use With (Application) | Quick PCR clean-up for downstream applications; Eliminates unincorporated primers and dNTPs |
| No. of Reactions | 2000 |
| Product Type | Single-step enzymatic PCR Cleanup Kit |
| Quantity | 4 x 1 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction