Learn More
Invitrogen™ Shrimp Alkaline Phosphatase (SAP)
Treat unincorporated dNTPs in PCR reactions to prepare templates for DNA sequencing or SNP analysis. Shrimp Alkaline Phosphatase (SAP) is useful in molecular biology applications such as the dephosphorylation of phosphorylated ends of DNA or RNA for subsequent use in cloning or end-labeling of probes.
£68.50 - £487.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15533687
|
Invitrogen™
78390500UN |
500 U |
£68.50
500 units |
Estimated Shipment: 13-12-2024
Log in to see stock. |
|||||
15523687
|
Invitrogen™
783901000UN |
1000 U |
£127.00
1000 units |
In Stock
Log in to see stock. |
|||||
15543687
|
Invitrogen™
783905000UN |
5000 U |
£487.00
5000 units |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
Description
- 100% heat-inactivated in 5 min at 65°C
- Significantly improved storage stability at lower temperatures
- Very high specific activity (see Fig. 3)
- Removes 5’-phosphates from DNA, RNA, dNTPs, and proteins
- Purified from a recombinant source
- May be added directly to restriction enzyme digests
- No vector purification necessary
- Requires no supplemental zinc or other additives for activity
- Works direct in many different buffers
- Easy treatment of unincorporated dNTPs in PCR products prior to DNA sequencing or SNP analysis
Specifications
Dephosphorylation of DNA/RNA |