Learn More
Terumo™ Agani™ Safety Needle
Safety hypodermic needle with secure safety activation and reliable sharps protection with one-hand activation.
Brand:  Terumo™ 8SAN1825R1
 View more versions of this product
Product Code. 17238066
  • £12.45 / Pack of 100
Explore more special offers
Description
Description
  • The Agani™ Safety hypodermic needles, when fitted with a standard luer slip or luer lock syringe are intended for the aspiration and injection of fluids for medical purposes.
  • After withdrawing the needle from the body, the attached needle safety shield can be activated to cover the needle immediately after use to minimize the risk of an accidental needle stick.
Specifications
Specifications
Not Autoclavable
18 G
25 mm
No
Pink
Yes
Single-use
Sterile