missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Agar, NF Grade

Powder, meets N.F. grade

£111.00 - £364.00

Specifications

Content And Storage 15°C to 30°C
Form Powder
Grade USP, NF
Product Type Agar
For Use With (Application) Used for nutrient media for bacterial cultures
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11462930
View Documents
MP Biomedicals™
0210026283
250 g
£119.00
250g
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
 
11452930
View Documents
MP Biomedicals™
0210026280
100 g
£111.00
100g
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
 
11472930
View Documents
MP Biomedicals™
0210026290
500 g
£233.00
500g
Estimated Shipment: 24-03-2025
to see stock.
Add to basket
 
11482930
View Documents
MP Biomedicals™
0210026291
1 kg
£364.00
1kg
Estimated Shipment: 24-03-2025
to see stock.
Add to basket
 
Description

Description

  • 80-100 mesh
  • Substitute for gelatin, isinglass
  • Used as nutrient media for bacterial culture
Specifications

Specifications

15°C to 30°C
USP, NF
Used for nutrient media for bacterial cultures
Powder
Agar
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ Agar, NF Grade

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.