missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Agar powder Product Code.: 11489632

Thermo Scientific Chemicals Agar powder

Product Code. 11489632
500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11489632

Brand: Thermo Scientific Chemicals A10752.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Agar is the most suited solidifying agent for scientific applications that require incubation temperatures closer to human body temperature. Agar is used as a medium in bacteriology and plant biology, impression material in dentistry, biofertilizer in organic farming, and salt bridges in electrochemistry. It can be used to measure the motility and mobility of microorganism owing to the porous nature of the gel form of an agar or agarose medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agar is the most suited solidifying agent for scientific applications that require incubation temperatures closer to human body temperature. Agar is used as a medium in bacteriology and plant biology, impression material in dentistry, biofertilizer in organic farming, and salt bridges in electrochemistry. It can be used to measure the motility and mobility of microorganism owing to the porous nature of the gel form of an agar or agarose medium.

Solubility
Soluble in water at temperatures between 950C and 1000 C. Insoluble in cold water.

Notes
Prolonged exposure to high temperatures can degrade Agar powder.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-18-0
Molecular Formula C14H24O9
Molecular Weight (g/mol) 336.337
MDL Number MFCD00081288
InChI Key GYYDPBCUIJTIBM-DYOGSRDZSA-N
Synonym agar, agar, pure, powder, agar agar bacteriological, 3r,4s,5s,6r-2-4r,5s-4-hydroxy-3-methyl-2,6-dioxabicyclo 3.2.1 octan-8-yl oxy-6-hydroxymethyl-4-methoxyoxane-3,5-diolShow MoreShow Less
PubChem CID 71571511
IUPAC Name (2R,3S,4S,5R)-2-(hydroxymethyl)-6-[[(4R,5S)-4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy]-4-methoxyoxane-3,5-diol
SMILES CC1C(C2C(C(O1)CO2)OC3C(C(C(C(O3)CO)O)OC)O)O

Specifications

Physical Form Powder
Quantity 500 g
Solubility Information Soluble in water at temperatures between 950C and 1000C. Insoluble in cold water.
Chemical Name or Material Agar

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN20L017 Certificate TypeCertificate of Analysis Date24/02/2025 Catalog NumberA10752.22,A10752.36,A10752.0E
Lot NumberX04K024 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog NumberA10752.22,A10752.36,A10752.0E
Lot Number10250630 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA10752.22,A10752.36,A10752.0E
Lot Number10251515 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA10752.22,A10752.36,A10752.0E
Lot NumberQ10K023 Certificate TypeCertificate of Analysis Date18/04/2024 Catalog NumberA10752.22,A10752.36,A10752.0E

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Agar powder >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.