missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Agarose
Agarose, Low Melting Point, Molecular Biology Grade, White Powder
Brand: MP Biomedicals™ 11AGAL0050
241.13 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Agarose low melting point is an intermediate melting and gelling point agarose which exhibits superior sieving characteristics.
- It is recommended for analytical gels of DNA fragments under 1000 bp and is ideal for small DNA fragments down to 20 bp can be separated on 6 % gels.
- It resolves fragments differing in size by only 2 bp.
- Its use avoids the need for PAGE Southern and Northern transfers can be achieved efficiently from them at concentrations up to 2%.
- DNA fragments can be purified with high efficiency from this agarose gels up to 4% by using either a purification kit or an electroelution system.
Specifications
9012-36-6 | |
Agarose | |
Nucleic Acidgel Electrophoresis & Blotting; Nucleic Acid Isolation & Purification; Nucleic Acid Electrophoresis; Nucleic Acid Sequencing | |
50 g |
White | |
Powder | |
< 67°C (1.5%gel) | |
Room Temperature |