missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™
£139.00 - £536.00
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10688973
|
Fisher BioReagents
BP1356-100 |
100 g | Poly Bottle |
£139.00
100g |
In Stock
Log in to see stock. |
|||||
10776644
|
Fisher BioReagents
BP1356-500 |
500 g | Poly Bottle |
£536.00
500g |
In Stock
Log in to see stock. |
|||||
Description
Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade)
Chemical Identifiers
9012-36-6 | |
C12H18O9 |
Specifications
Powder/Solid | |
Cream | |
0.15 max. | |
1200g/cm2min. | |
306.12 | |
Agarose, Broad Separation Range for DNA/RNA (500bp to 25kb) |
9012-36-6 | |
DNase free | |
34.5° to 37.5°C | |
C12H18O9 | |
DNase- and RNase-Free |
Safety and Handling
Recommended Storage : RT
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™