missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Agarose, Electrophoresis Grade Product Code.: 15427889

Thermo Scientific Chemicals Agarose, Electrophoresis Grade

Product Code. 15427889
50 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 15427889

Brand: Thermo Scientific Chemicals J66501.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Standard agarose suitable for most general applications. Good gel strength and clarity. Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Standard agarose suitable for most general applications. Good gel strength and clarity. Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it.

Solubility
Soluble in water(10g/L).

Notes
Gel strength (1.5% w/v): >1120g/cm^2; Gelling temperature: 36-39°C; EEO: <0.12
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Physical Form Powder
Melting Point 87°C to 89°C
Color White
Quantity 50 g
Sensitivity Hygroscopic; Moisture sensitive
Solubility Information Soluble in water(10g/L).
Grade Electrophoresis
Chemical Name or Material Agarose

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN14L557 Certificate TypeCertificate of Analysis Date22/02/2025 Catalog NumberJ66501.30,J66501.18
Lot NumberY26K568 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberJ66501.30,J66501.18
Lot NumberX29K524 Certificate TypeCertificate of Analysis Date07/11/2024 Catalog NumberJ66501.30,J66501.18
Lot NumberW16K500 Certificate TypeCertificate of Analysis Date23/09/2024 Catalog NumberJ66501.30,J66501.18
Lot NumberU25K521 Certificate TypeCertificate of Analysis Date29/08/2024 Catalog NumberJ66501.30,J66501.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Agarose, Electrophoresis Grade >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.